missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GCDH Protein
A cDNA sequence encoding the GCDH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11938-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
2639 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRPEFDWQDP LVLEEQLTTD EILIRDTFRT YCQERLMPRI LLANRNEVFH REIISEMGEL GVLGPTIKGY GCAGVSSVAY GLLARELERV DSGYRSAMSV QSSLVMHPIY AYGSEEQRQK YLPQLAKGEL LGCFGLTEPN SGSDPSSMET RAHYNSSNKS YTLNGTKTWI TNSPMADLFV VWARCEDGCI RGFLLEKGMR GLSAPRIQGK FSLRASATGM IIMDGVEVPE ENVLPGASSL GGPFGCLNNA RYGIAWGVLG ASEFCLHTAR QYALDRMQFG VPLARNQLIQ KKLADMLTEI TLGLHACLQL GRLKDQDKAA PEMVSLLKRN NCGKALDIAR QARDMLGGNG ISDEYHVIRH AMNLEAVNTY EGTHDIHALI LGRAITGIQA FTASK | |
Greater than 90.0% as determined by SDS-PAGE. |
GCDH Protein | |
Research Use Only | |
GCDH | |
Human | |
His | |
0.5 mg/ml solution containing 20mM Tris-HCl, pH 8.0, 5mM DTT, 0.2M NaCl and 20% glycerol. |