missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GDF15 D Protein

A cDNA sequence encoding the GDF15 D was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11949-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15938898

  • 4385.00€ / 1mg


Spedizione stimata
11-01-2021
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

GDF15 D Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
GDF15 D-variant is lyophilized without additives.
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MARNGDDCPL GPGRCCRLHT VRASLEDLGW ADWVLSPREV QVTMCIGACP SQFRAANMHA QIKTSLHRLK PDTVPAPCCV PASYNPMVLI QKTDTGVSLQ TYDDLLAKDC HCI
Videos
SDS
Documenti