missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Glycerol kinase 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Glycerol kinase 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6085-ec-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15939859

Quantità Prezzo
1 675.00€ / 200µg
5 629.51€ / 200µg
Spedizione stimata
14-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_149991.2
Glycerol kinase 2 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2712
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GK2
Recombinant Protein
E. coli
AAPKTAAVGP LVGAVVQGTN STRFLVFNSK TAELLSHHKV ELTQEFPKEG WVEQDPKEIL QSVYECIART CEKLDELNID ISNIKAVGVS NQRETTVIWD KLTGEPLYNA VVWLDLRTQT TVEDLSKKIP GNSNFVKSKT GLPLSTYFSA VKLRWMLDNV RNVQKAVEEG RALFGTIDSW LIWSLTGGVN GGVHCTDVTN ASRTMLFNIH SLEWDKELCD FFEIPMDLLP NVFSSSEIYG LIKTGALEGV PISGCLGDQC AALVGQMCFQ EGQAKNTYGT GCFLLCNTGR KCVFSEHGLL TTVAYKLGRE KPAYYALEGS VAIAGAVIRW LRDNLGIIET SGDIERLAKE VGTSYGCYFV PAFSGLYAPY WEPSARGILC GLTQFTNKCH IAFAALEAVC FQTREILEAM NRDCGIPLRH LQVDGGMTNN KVLMQLQADI LHIPVIKPFM PETTALGAAM AAGAAEGVSV WSLEPQALSV LRMERFEPQI QATESEIRYA TWKKAVMKSM GWVTSQSPEG GDPSIFSSLP LGFFIVSSMV MLIGARYISG VP
Videos
SDS
Documenti