missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GNB1L Protein
A DNA sequence encoding the Homo sapiens (Human) GNB1L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8085-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_443730.1 | |
GNB1L Protein | |
Research Use Only | |
GNB1L | |
Recombinant Protein | |
E. coli | |
MTAPCPPPPP DPQFVLRGTQ SPVHALHFCE GAQAQGRPLL FSGSQSGLVH IWSLQTRRAV TTLDGHGGQC VTWLQTLPQG RQLLSQGRDL KLCLWDLAEG RSAVVDSVCL ESVGFCRSSI LAGGQPRWTL AVPGRGSDEV QILEMPSKTS VCALKPKADA KLGMPMCLRL WQADCSSRPL LLAGYEDGSV VLWDVSEQKV CSRIACHEEP VMDLDFDSQK ARGISGSAGK ALAVWSLDWQ QALQVRGTHE LTNPGIAEVT IRPDRKILAT AGWDHRIRVF HWRTMQPLAV LAFHSAAVQC VAFTADGLLA AGSKDQRISL WSLYPRA | |
Greater than 80% as determined by SDS-PAGE. |
54584 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |