missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GPNMB Protein

A cDNA sequence encoding the GPNMB was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12041-HIS-HEK-2ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15949178

  • 137.00€ / 2µg
Spedizione stimata: 18-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

GPNMB Protein
Research Use Only
Recombinant Protein
Human (HEK293)
AKRFHDVLGN ERPSAYMREH NQLNGWSSDE NDWNEKLYPV WKRGDMRWKN SWKGGRVQAV LTSDSPALVG SNITFAVNLI FPRCQKEDAN GNIVYEKNCR NEAGLSADPY VYNWTAWSED SDGENGTGQS HHNVFPDGKP FPHHPGWRRW NFIYVFHTLG QYFQKLGRCS VRVSVNTANV TLGPQLMEVT VYRRHGRAYV PIAQVKDVYV VTDQIPVFVT MFQKNDRNSS DETFLKDLPI MFDVLIHDPS HFLNYSTINY KWSFGDNTGL FVSTNHTVNH TYVLNGTFSL NLTVKAAAPG PCPPPPPPPR PSKPTPSLAT TLKSYDSNTP GPAGDNPLEL SRIPDENCQI NRYGHFQATI TIVEGILEVN IIQMTDVLMP VPWPESSLID FVVTCQGSIP TEVCTIISDP TCEITQNTVC SPVDVDEMCL LTVRRTFNGS GTYCVNLTLG DDTSLALTST LISVP KLHHH HHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl, pH 7.4.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali