missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HDL Protein
A cDNA sequence encoding the HDL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12211-1gram
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
HDL Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
Cholesterol level >200 mg/l. <1% LDL. |
1 g | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged |