missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2A type 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2A type 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6158-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_003500.1 | |
Histone H2A type 1 Protein | |
Research Use Only | |
HIST1H2AG | |
Recombinant Protein | |
E. coli | |
SGRGKQGGKA RAKAKTRSSR AGLQFPVGRV HRLLRKGNYA ERVGAGAPVY LAAVLEYLTA EILELAGNAA RDNKKTRIIP RHLQLAIRND EELNKLLGKV TIAQGGVLPN IQAVLLPKKT ESHHKAKGK | |
Greater than 90% as determined by SDS-PAGE. |
8329 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |