missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2A.V Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2A.V, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7750-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036544.1 | |
Histone H2A.V Protein | |
Research Use Only | |
H2AFV | |
Recombinant Protein | |
E. coli | |
MAGGKAGKDS GKAKAKAVSR SQRAGLQFPV GRIHRHLKTR TTSHGRVGAT AAVYSAAILE YLTAEVLELA GNASKDLKVK RITPRHLQLA IRGDEELDSL IKATIAGGGV IPHIHKSLIG KKGQQKTA | |
Greater than 90% as determined by SDS-PAGE. |
94239 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |