missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGF1 Isoform 2 Protein

A DNA sequence encoding the Human IGF1 Isoform 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10390-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943708

Quantità Prezzo
1 105.00€ / 10µg
10 98.00€ / 10µg
Spedizione stimata
18-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_000609.1
IGF1 Isoform 2 Protein
10 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
3479
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
IGF1
The ED50 as determined by a cell proliferation assay using serum free human MCF-7 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.0, lyophilized
Videos
SDS
Documenti