missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-1RA / IL1RN Protein
A DNA sequence encoding the Human IL-1RA / IL1RN, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10308-ec-250ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000568.1 | |
IL-1RA / IL1RN Protein | |
Research Use Only | |
IL1RN | |
The ED50 as determined by inhibiting IL-1alpha- dependent proliferation of murine D10S cells is less than 40 ng/ml, corresponding to a specific activity of > 2.5 M 104 IU/mg in the presence of 50 pg/ml rHuIL-1alpha. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3557 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
RPSGRKSSKM QAFRIWDVNQ KTFYLRNNQL VAGYLQGPNV NLEEKIDVVP IEPHALFLGI HGGKMCLSCV KSGDETRLQL EAVNITDLSE NRKQDKRFAF IRSDSGPTTS FESAACPGWF LCTAMEADQP VSLTNMPDEG VMVTKFYFQE DE | |
Greater than 96% as determined by SDS-PAGE and HPLC. |