missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-6R alpha Fc Chimera Protein

Product Code. 18796315
Change view
Click to view available options
Quantità:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
18796315 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796315 Supplier R&D Systems™ Supplier No. 11399SR050
 Articolo soggetto a restrizioni
Spedizione stimata: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its binding ability in a functional ELISA. Recombinant Human IL-6R alpha Fc Chimera (Catalog # 11399-SR) binds Biotinylated Recombinant Human IL-6 (Catalog # BT7270 ) with an ED50 of 0.750-7.50 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Numero di accesso P08887.1
Da utilizzare con (applicazione) Bioactivity
Formulazione Supplied as a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details.
ID gene (immissione) 3570
Peso molecolare MolecularWeight-observed: 88-99 kDa, under reducing conditions., MolecularWeight-theroretical: 65 kDa
Quantità 50 μg
Sorgente Chinese Hamster Ovary cell line, CHO-derived human IL-6 R alpha protein Human IL-6 R alpha (Leu20-Pro365) (N-terminus) Accession # P08887.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Requisiti di stoccaggio Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Concentrazione di endotossine <0.10 EU / 1 μg of the protein by the LAL method.
Alias gene CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6 R-1, IL6RA, IL-6 RA, IL6RQ, interleukin 6 receptor
Coniugato Unconjugated
Linea di prodotti R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Proteine IL-6 R alpha
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.