Learn More
enQuireBio™ Recombinant Human IL33 / Interleukin-33 / NF-HEV Protein
Recombinant Protein
Marca:  enQuireBio™ QP1168-1 mg
Dettagli aggiuntivi : Peso : 0.01000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15938018
  • 5780.00€ / 1mg
Spedizione stimata
08-01-2021
Aggiungi al carrello
Specifica
NP_001186569.1
IL33 / Interleukin-33 / NF-HEV Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 500 pg/mL in the D10S cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^6 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5
90865
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL33
Recombinant Protein
E. coli
MSITGISPIT EYLASLSTYN DQSITFALED ESYEIYVEDL KKDEKKDKVL LSYYESQHPS NESGDGVDGK MLMVTLSPTK DFWLHANNKE HSVELHKCEK PLPDQAFFVL HNMHSNCVSF ECKTDPGVFI GVKDNHLALI KVDSSENLCT ENILFKLSET