missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha 2b Protein
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha 2b, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7345-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000596.2 | |
Interferon alpha 2b Protein | |
Research Use Only | |
IFNA2 | |
Recombinant Protein | |
E. coli | |
CDLPQTHSLG SRRTLMLLAQ MRKISLFSCL KDRHDFGFPQ EEFGNQFQKA ETIPVLHEMI QQIFNLFSTK DSSAAWDETL LDKFYTELYQ QLNDLEACVI QGVGVTETPL MKEDSILAVR KYFQRITLYL KEKKYSPCAW EVVRAEIMRS FSLSTNLQES LRSKE | |
Greater than 90% as determined by SDS-PAGE. |
3440 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |