missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IZUMO4 / C19orf36 Protein

A DNA sequence encoding the Homo sapiens (Human) IZUMO4 / C19orf36, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7632-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16059910

Quantità Prezzo
1 1855.00€ / 1mg
3 1731.09€ / 1mg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001026905.2
IZUMO4 / C19orf36 Protein
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
113177
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IZUMO4
Recombinant Protein
E. coli
HGCLHCHSNF SKKFSFYRHH VNFKSWWVGD IPVSGALLTD WSDDTMKELH LAIPAKITRE KLDQVATAVY QMMDQLYQGK MYFPGYFPNE LRNIFREQVH LIQNAIIESR IDCQHRCGIF QYETISCNNC TDSHVACFGY NCESSAQWKS AVQGLLNYIN NWHKQDTSMS LVSPALRCLE PPHLANLTLE DAAECLKQH
Videos
Documenti