Learn More
enQuireBio™ Recombinant Human KLF6 Protein
A cDNA sequence encoding the KLF6 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifica
Nome KLF6 Protein
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Human
Mehr Spezifikationen ansehen
Products 3
Produktcode Marke Quantità Preis Menge & Verfügbarkeit  
Produktcode Marke Quantità Preis Menge & Verfügbarkeit  
15910619
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP12499-2UG
2 μg
137.00€
2µg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
15990609
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP12499-10UG
10 μg
212.00€
10µg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
15900619
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP12499-1MG
1 mg
7875.00€
1mg
Alternative Produkte anzeigen
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
Beschreibung
Beschreibung
Spezifikation
Spezifikation
KLF6 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The KLF6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMDVLPMC SIFQELQIVH ETGYFSALPS LEEYWQQTCL ELERYLQSEP CYVSASEIKF DSQEDLWTKI ILAREKKEES ELKISSSPPE DTLISPSFCY NLETNSLNSD VSSESSDSSE ELSPTAKFTS DPIGEVLVSS GKLSSSVTST PPSSPELSRE PSQLWGCVPG ELPSPGKVRS GTSGKPGDKG NGDASPDGRR RVHRCHFNGC RKVYTKSSHL KAHQRTHTGE KPYRCSWEGC EWRFARSDEL TRHFRKHTGA KPFKCSHCDR CFSRSDHLAL HMKRHL
Greater than 85% as determined by SDS-PAGE.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate