missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kynurenine 3-monooxygenase Protein

A DNA sequence encoding the Homo sapiens (Human) Kynurenine 3-monooxygenase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6271-ec-50ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062090

Quantità Prezzo
1 234.00€ / 50µg
6 218.51€ / 50µg
Spedizione stimata
28-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_003670.2
Kynurenine 3-monooxygenase Protein
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8564
Greater than 90% as determined by SDS-PAGE.
Research Use Only
KMO
Recombinant Protein
E. coli
MDSSVIQRKK VAVIGGGLVG SLQACFLAKR NFQIDVYEAR EDTRVATFTR GRSINLALSH RGRQALKAVG LEDQIVSQGI PMRARMIHSL SGKKSAIPYG TKSQYILSVS RENLNKDLLT AAEKYPNVKM HFNHRLLKCN PEEGMITVLG SDKVPKDVTC DLIVGCDGAY STVRSHLMKK PRFDYSQQYI PHGYMELTIP PKNGDYAMEP NYLHIWPRNT FMMIALPNMN KSFTCTLFMP FEEFEKLLTS NDVVDFFQKY FPDAIPLIGE KLLVQDFFLL PAQPMISVKC SSFHFKSHCV LLGDAAHAIV PFFGQGMNAG FEDCLVFDEL MDKFSNDLSL CLPVFSRLRI PDDHAISDLS MYNYIEMRAH VNSSWFIFQK NMERFLHAIM PSTFIPLYTM VTFSRIRYHE AVQRWHWQKK VINKGLFFLG SLIAISSTYL LIHYMSPRSF LRLRRPWNWI AHFRNTTCFP AKAVDSLEQI SNLISR
Videos
Documenti