missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lck Kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Lck Kinase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6288-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001036236.1 | |
Lck Kinase Protein | |
10 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3932 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LCK | |
Recombinant Protein | |
E. coli | |
MGCGCSSHPE DDWMENIDVC ENCHYPIVPL DGKGTLLIRN GSEVRDPLVT YEGSNPPASP LQDNLVIALH SYEPSHDGDL GFEKGEQLRI LEQSGEWWKA QSLTTGQEGF IPFNFVAKAN SLEPEPWFFK NLSRKDAERQ LLAPGNTHGS FLIRESESTA GSFSLSVRDF DQNQGEVVKH YKIRNLDNGG FYISPRITFP GLHELVRHYT NASDGLCTRL SRPCQTQKPQ KPWWEDEWEV PRETLKLVER LGAGQFGEVW MGYYNGHTKV AVKSLKQGSM SPDAFLAEAN LMKQLQHQRL VRLYAVVTQE PIYIITEYME NDTLLDSQLE EKGLGASPWG NLGQQLLLLP TGSLVDFLKT PSGIKLTINK LLDMAAQIAE GMAFIEERNY IHRDLRAANI LVSDTLSCKI ADFGLARLIE DNEYTAREGA KFPIKWTAPE AINYGTFTIK SDVWSFGILL TEIVTHGRIP YPGMTNPEVI QNLERGYRMV RPDNCPEELY QLMRLCWKER PEDRPTFDYL RSVLEDFFTA TEGQYQPQP |