Learn More
enQuireBio™ Recombinant Human Legumain / LGMN Protein
Codice prodotto. 16082290
Cambia vista
Click to view available options
Quantità:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Dimensione della confezione:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16082290 100 μg 100µg
16092290 10 μg 10µg
16032300 50 μg 50µg
16012300 200 μg 200µg
16022300 500 μg 500µg
16002300 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 16082290 Fornitore enQuireBio™ N. del fornitore QP6301ec100ug
Please to purchase this item. Need a web account? Register with us today!
A DNA sequence encoding the Homo sapiens (Human) Legumain/ LGMN, was expressed in the hosts and tags indicated.
Specifica
Numero di accesso NP_001008530.1
ID gene (immissione) 5641
Nome Legumain / LGMN Protein
Quantità 100 μg
Status giuridico Research Use Only
Concentrazione di endotossine Not determined.
Simbolo del gene Lgmn
Lunghezza della proteina Full Length of Mature Protein
Product Type Recombinant Protein
Cross-reattività Human
Specie E. coli
Tag proteine His
Sequenza VPIDDPEDGG KHWVVIVAGS NGWYNYRHQA DACHAYQIIH RNGIPDEQIV VMMYDDIAYS EDNPTPGIVI NRPNGTDVYQ GVPKDYTGED VTPQNFLAVL RGDAEAVKGI GSGKVLKSGP QDHVFIYFTD HGSTGILVFP NEDLHVKDLN ETIHYMYKHK MYRKMVFYIE ACESGSMMNH LPDNINVYAT TAANPRESSY ACYYDEKRST YLGDWYSVNW MEDSDVEDLT KETLHKQYHL VKSHTNTSHV MQYGNKTIST MKVMQFQGMK RKASSPVPLP PVTHLDLTPS PDVPLTIMKR KLMNTN
Tampone Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.