missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIPA Protein

A DNA sequence encoding the Homo sapiens (Human) LIPA, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6311-ye-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16012390

Quantità Prezzo
1 524.24€ / 100µg
5 493.36€ / 100µg
Spedizione stimata
01-06-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_000226.2
LIPA Protein
100 μg
Not determined.
Full Length of Mature Protein
Human
His
Tris-based buffer, 50% glycerol
3988
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LIPA
Recombinant Protein
Yeast
SGGKLTAVDP ETNMNVSEII SYWGFPSEEY LVETEDGYIL CLNRIPHGRK NHSDKGPKPV VFLQHGLLAD SSNWVTNLAN SSLGFILADA GFDVWMGNSR GNTWSRKHKT LSVSQDEFWA FSYDEMAKYD LPASINFILN KTGQEQVYYV GHSQGTTIGF IAFSQIPELA KRIKMFFALG PVASVAFCTS PMAKLGRLPD HLIKDLFGDK EFLPQSAFLK WLGTHVCTHV ILKELCGNLC FLLCGFNERN LNMSRVDVYT THSPAGTSVQ NMLHWSQAVK FQKFQAFDWG SSAKNYFHYN QSYPPTYNVK DMLVPTAVWS GGHDWLADVY DVNILLTQIT NLVFHESIPE WEHLDFIWGL DAPWRLYNKI INLMRKYQ
Videos
SDS
Documenti