missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LRP4 Protein

A DNA sequence encoding the Homo sapiens (Human) LRP4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6320-ec-50ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062450

Quantità Prezzo
1 234.00€ / 50µg
6 218.51€ / 50µg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_002325.2
LRP4 Protein
50 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
4038
Greater than 95% as determined by SDS-PAGE.
Research Use Only
LRP4
Recombinant Protein
E. coli
YRHKKSKFTD PGMGNLTYSN PSYRTSTQEV KIEAIPKPAM YNQLCYKKEG GPDHNYTKEK IKIVEGICLL SGDDAEWDDL KQLRSSRGGL LRDHVCMKTD TVSIQASSGS LDDTETEQLL QEEQSECSSV HTAATPERRG SLPDTGWKHE RKLSSESQV
Videos
Documenti