missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LY-96 / ESOP-1 Protein
A DNA sequence encoding the Homo sapiens (Human) LY-96 / ESOP-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6326-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001182726.1 | |
LY-96 / ESOP-1 Protein | |
Research Use Only | |
LY96 | |
Recombinant Protein | |
E. coli | |
EAQKQYWVCN SSDASISYTY CDKMQYPISI NVNPCIELKR SKGLLHIFYI PRRDLKQLYF NLYITVNTMN LPKRKEVICR GSDDDYSFCR ALKGETVNTT ISFSFKGIKF SKGKYKCVVE AISGSPEEML FCLEFVILHQ PNSN | |
Greater than 80% as determined by SDS-PAGE. |
23643 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-GST | |
Tris-based buffer, 50% glycerol |