missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAP1LC3B2 Protein

A cDNA sequence encoding the MAP1LC3B2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12637-20ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15941069

Quantità Prezzo
1 166.00€ / 20µg
10 155.97€ / 20µg
Spedizione stimata
04-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

MAP1LC3B2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MAP1LC3B2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MPSEKTFKQR RTFEQRVEDV RLIREQHPTK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELIKIIRRR LQLNANQAFF LLVNGHSMVS VSTPISEVYE SEKDEDGFLY MVCASQETFG
Videos
SDS
Documenti