missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mast cell carboxypeptidase A Protein

A DNA sequence encoding the Homo sapiens (Human) Mast cell carboxypeptidase A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5856-ye-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15938269

Quantità Prezzo
1 805.00€ / 200µg
5 750.50€ / 200µg
Spedizione stimata
14-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_001861.2
Mast cell carboxypeptidase A Protein
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
1359
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CPA3
Recombinant Protein
Yeast
IPGRHSYAKY NNWEKIVAWT EKMMDKYPEM VSRIKIGSTV EDNPLYVLKI GEKNERRKAI FTDCGIHARE WVSPAFCQWF VYQATKTYGR NKIMTKLLDR MNFYILPVFN VDGYIWSWTK NRMWRKNRSK NQNSKCIGTD LNRNFNASWN SIPNTNDPCA DNYRGSAPES EKETKAVTNF IRSHLNEIKV YITFHSYSQM LLFPYGYTSK LPPNHEDLAK VAKIGTDVLS TRYETRYIYG PIESTIYPIS GSSLDWAYDL GIKHTFAFEL RDKGKFGFLL PESRIKPTCR ETMLAVKFIA KYILKHTS
Videos
SDS
Documenti