missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MEIS3 Protein
A cDNA sequence encoding the MEIS3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12675-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MEIS3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMARRYDE LPHYPGIVDG PAALASFPET VPAVPGPYGP HRPPQPLPPG LDSDGLKREK DEIYGHPLFP LLALVFEKCE LATCSPRDGA GAGLGTPPGG DVCSSDSFNE DIAAFAKQVR SERPLFSSNP ELDNLMIQAI QVLRFHLLEL EKGKMPIDLVIEDRDGGCRE DFEDYPASCP SLPDQNNMWI RDHEDSGSVH LGTPGPSSGG LASQSGDNSS DQGDGLDTSV ASPSSGGEDE DLDQERRRNK KRGIFPKVAT NIMRAWLFQH LSHPYPSEEQ KKQLAQDTGL TILQVNNWFI NARRRIVQPM IDQSNRTGQG AAFSPEGQPI GGYTETQPHVAVRPPGSVGM SLNLEGEWHY L | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MEIS3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |