missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MEP1A Protein
A cDNA sequence encoding the MEP1A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12678-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MEP1A Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPVPIKYLP EENVHDADFG EQKDISEINL AAGLDLFQGD ILLQKSRNGL RDPNTRWTFP IPYILADNLG LNAKGAILYA FEMFRLKSCV DFKPYEGESS YIIFQQFDGC WSEVGDQHVG QNISIGQGCA YKAIIEHEIL HALGFYHEQS RTDRDDYVNI WWDQILSGYQ HNFDTYDDSL ITDLNTPYDY ESLMHYQPFS FNKNASVPTI TAKIPEFNSI IGQRLDFSAI DLERLNRMYN CTTTHTLLDH CTFEKANICG MIQGTRDDTD WAHQDSAQAG EVDHTLLGQC TGAGYFMQFS TSSGSAEEAA LLESRILYPK RKQQCLQFFY KMTGSPSDRL VVWVRRDDST GNVRKLVKVQ TFQGDDDHNW KIAHVVLKEE QKFRYLFQGT KGDPQNSTGG IYLDDITLTE TPCPTGVWTV RNFSQVLENT SKGDKLQSPR FYNSEGYGFG VTLYPNSRES SGYLRLAFHV CSGENDAILE WPVENRQVII TILDQEPDVR NRMSSSMVFT TSKSHTSPAI NDTVIWDRPS RVGTYHTDCN CFRSIDLGWS GFISHQMLKR RSFLKNDDLI IFVDFEDITH LSQHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MEP1A protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |