missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Meteorin-like Protein
A DNA sequence encoding the Homo sapiens (Human) Meteorin-like protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7719-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001004431.1 | |
Meteorin-like Protein | |
Research Use Only | |
METRNL | |
Recombinant Protein | |
Yeast | |
QYSSDRCSWK GSGLTHEAHR KEVEQVYLRC AAGAVEWMYP TGALIVNLRP NTFSPARHLT VCIRSFTDSS GANIYLEKTG ELRLLVPDGD GRPGRVQCFG LEQGGLFVEA TPQQDIGRRT TGFQYELVRR HRASDLHELS APCRPCSDTE VLLAVCTSDF AVRGSIQQVT HEPERQDSAI HLRVSRLYRQ KSRVFEPVPE GDGHWQGRVR TLLECGVRPG HGDFLFTGHM HFGEARLGCA PRFKDFQRMY RDAQERGLNP CEVGTD | |
Greater than 90% as determined by SDS-PAGE. |
284207 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |