missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MKK6 / MEK6 / MAP2K6 Protein

A DNA sequence encoding the Homo sapiens (Human) MKK6 / MEK6 / MAP2K6, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6338-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062580

Quantità Prezzo
1 406.00€ / 100µg
6 379.00€ / 100µg
Spedizione stimata
21-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_002749.2
MKK6 / MEK6 / MAP2K6 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5608
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MAP2K6
Recombinant Protein
E. coli
MSQSKGKKRN PGLKIPKEAF EQPQTSSTPP RDLDSKACIS IGNQNFEVKA DDLEPIMELG RGAYGVVEKM RHVPSGQIMA VKRIRATVNS QEQKRLLMDL DISMRTVDCP FTVTFYGALF REGDVWICME LMDTSLDKFY KQVIDKGQTI PEDILGKIAV SIVKALEHLH SKLSVIHRDV KPSNVLINAL GQVKMCDFGI SGYLVDSVAK TIDAGCKPYM APERINPELN QKGYSVKSDI WSLGITMIEL AILRFPYDSW GTPFQQLKQV VEEPSPQLPA DKFSAEFVDF TSQCLKKNSK ERPTYPELMQ HPFFTLHESK GTDVASFVKL ILGD
Videos
Documenti