Learn More
R&D Systems™ Recombinant Human/Mouse/Rat GDF-8/Myostatin Protein, CF
Product Code. 30306761
Change view
Click to view available options
Quantity:
100 μg
20 μg
250 μg
50 μg
Unit Size:
100µg
20µg
250µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30306761 50 μg 50µg
30306759 20 μg 20µg
30306772 100 μg 100µg
30306754 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30306761 Supplier R&D Systems™ Supplier No. 11658G8050
 Restricted Item
Spedizione stimata: 02-12-2026
Recombinant Protein
Measured by its ability to induce cytotoxicity using MPC-11 mouse B lymphocyte cells. The ED50 for this effect is 4.00-40.0 ng/mL.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number O14793.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 2660
Molecular Weight (g/mol) MolecularWeight-observed: 10-14 kDa, under reducing conditions., MolecularWeight-theroretical: 12 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived GDF-8/Myostatin protein Asp267-Ser375
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Conjugate Unconjugated
Reconstitution Reconstitute at 250 μg/mL in water.
Product Line R&D Systems
Protein GDF-8/Myostatin