missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MXD3 Protein
A cDNA sequence encoding the MXD3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12764-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
MXD3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEPLASN IQVLLQAAEF LERREREAEH GYASLCPHRS PGPIHRRKKR PPQAPGAQDS GRSVHNELEK RRRAQLKRCL ERLKQQMPLG ADCARYTTLS LLRRARMHIQ KLEDQEQRAR QLKERLRSKQ QSLQRQLEQL RGLAGAAERE RLRADSLDSS GLSSERSDSD QEELEVDVES LVFGGEAELL RGFVAGQEHS YSHGGGAWL | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MXD3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |