missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N6AMT1 / HEMK2 Protein
A DNA sequence encoding the Homo sapiens (Human) N6AMT1 / HEMK2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6156-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_037372.4 | |
N6AMT1 / HEMK2 Protein | |
Research Use Only | |
N6AMT1 | |
Recombinant Protein | |
E. coli | |
MAGENFATPF HGHVGRGAFS DVYEPAEDTF LLLNALEAAA AELAGVEICL EVGSGSGVVS AFLASMIGPQ ALYMCTDINP EAAACTLETA RCNKVHIQPV ITDLVGSHGI EAAWAGGKNG REVMDRFFPL VPDLLSPKGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFTKS | |
Greater than 80% as determined by SDS-PAGE. |
29104 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |