missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Neurotrypsin Protein

A DNA sequence encoding the Homo sapiens (Human) Neurotrypsin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6537-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16014020

Quantità Prezzo
1 2825.92€ / 1mg
3 2658.91€ / 1mg
Spedizione stimata
07-06-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_003610.2
Neurotrypsin Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
8492
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PRSS12
Recombinant Protein
E. coli
IIGGKNSLRG GWPWQVSLRL KSSHGDGRLL CGATLLSSCW VLTAAHCFKR YGNSTRSYAV RVGDYHTLVP EEFEEEIGVQ QIVIHREYRP DRSDYDIALV RLQGPEEQCA RFSSHVLPAC LPLWRERPQK TASNCYITGW GDTGRAYSRT LQQAAIPLLP KRFCEERYKG RFTGRMLCAG NLHEHKRVDS CQGDSGGPLM CERPGESWVV YGVTSWGYGC GVKDSPGVYT KVSAFVPWIK SVTK
Videos
SDS
Documenti