missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NFASC / Neurofascin Protein

A DNA sequence encoding the Homo sapiens (Human) NFASC / Neurofascin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6416-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16013160

Quantità Prezzo
1 1979.08€ / 1mg
3 1861.92€ / 1mg
Spedizione stimata
09-06-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_001005388.2
NFASC / Neurofascin Protein
1 mg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
23114
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NFASC
Recombinant Protein
E. coli
KRSRGGKYPV REKKDVPLGP EDPKEEDGSF DYSDEDNKPL QGSQTSLDGT IKQQESDDSL VDYGEGGEGQ FNEDGSFIGQ YTVKKDKEET EGNESSEATS PVNAIYSLA
Videos
SDS
Documenti