missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Non-secretory ribonuclease Protein
A DNA sequence encoding the Homo sapiens (Human) Non-secretory ribonuclease, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6610-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002925.1 | |
Non-secretory ribonuclease Protein | |
Research Use Only | |
RNASE2 | |
Recombinant Protein | |
E. coli | |
KPPQFTWAQW FETQHINMTS QQCTNAMQVI NNYQRRCKNQ NTFLLTTFAN VVNVCGNPNM TCPSNKTRKN CHHSGSQVPL IHCNLTTPSP QNISNCRYAQ TPANMFYIVA CDNRDQRRDP PQYPVVPVHL DRII | |
Greater than 90% as determined by SDS-PAGE. |
6036 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |