missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NR2F6 Protein

A DNA sequence encoding the Homo sapiens (Human) NR2F6, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6434-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16063280

Quantità Prezzo
1 245.00€ / 10µg
6 228.51€ / 10µg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_005225.2
NR2F6 Protein
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2063
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NR2F6
Recombinant Protein
E. coli
MAMVTGGWGG PGGDTNGVDK AGGYPRAAED DSASPPGAAS DAEPGDEERP GLQVDCVVCG DKSSGKHYGV FTCEGCKSFF KRSIRRNLSY TCRSNRDCQI DQHHRNQCQY CRLKKCFRVG MRKEAVQRGR IPHSLPGAVA ASSGSPPGSA LAAVASGGDL FPGQPVSELI AQLLRAEPYP AAAGRFGAGG GAAGAVLGID NVCELAARLL FSTVEWARHA PFFPELPVAD QVALLRLSWS ELFVLNAAQA ALPLHTAPLL AAAGLHAAPM AAERAVAFMD QVRAFQEQVD KLGRLQVDSA EYGCLKAIAL FTPDACGLSD PAHVESLQEK AQVALTEYVR AQYPSQPQRF GRLLLRLPAL RAVPASLISQ LFFMRLVGKT PIETLIRDML LSGSTFNWPY GSGQ
Videos
Documenti