missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human p38 delta / MAPK13 Protein

A DNA sequence encoding the Homo sapiens (Human) p38 delta / MAPK13, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6339-ec-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062590

Quantità Prezzo
1 1180.00€ / 500µg
3 1100.47€ / 500µg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_002745.1
p38 delta / MAPK13 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5603
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MAPK13
Recombinant Protein
E. coli
MSLIRKKGFY KQDVNKTAWE LPKTYVSPTH VGSGAYGSVC SAIDKRSGEK VAIKKLSRPF QSEIFAKRAY RELLLLKHMQ HENVIGLLDV FTPASSLRNF YDFYLVMPFM QTDLQKIMGM EFSEEKIQYL VYQMLKGLKY IHSAGVVHRD LKPGNLAVNE DCELKILDFG LARHADAEMT GYVVTRWYRA PEVILSWMHY NQTVDIWSVG CIMAEMLTGK TLFKGKDYLD QLTQILKVTG VPGTEFVQKL NDKAAKSYIQ SLPQTPRKDF TQLFPRASPQ AADLLEKMLE LDVDKRLTAA QALTHPFFEP FRDPEEETEA QQPFDDSLEH EKLTVDEWKQ HIYKEIVNFS PIARKDSRRR SGMKL
Videos
Documenti