missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PAPP A Protein

A cDNA sequence encoding the PAPP A was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12957-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15952069

  • 181.00€ / 10µg

Spedizione stimata: 07-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Specifica

Specifica

PAPP A Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PAPP-A protein is supplied in 1x PBS and 50% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRLWSWVLHL GLLSAALGCG LAERPRRARR DPRAGRPPRP AAGPATCATR AARGRRASPP PPPPPGGAWE AVRVPRRRQQ REARGATEEP SPPSRALYFS GRGEQLRLRA DLELPRDAFT LQVWLRAEGG QRSPAVITGL YDKCSYISRD RGWVVGIHTI SDQDNKDPRY FFSLKTDRAR QVTTINAHRS YLPGQWVYLA ATYDGQFMKL YVNGAQVATS GEQVGGIFSP LTQKCKVLML GGSALNHNYR GYIEHFSLWK VARTQREILS DMETHGAHTA LPQLLLQENW DNVKHAWSPM KDGSSPKVEF SNAHGFLLDT SLEPPLCGQT LCDNTEVIAS YNQLSSFRQP KVVRYRVVNL YEDDHKNPTV TREQVDFQHH QLAEAFKQYN ISWELDVLEV SNSSLRRRLI LANCDISKIG DENCDPECNH TLTGHDGGDC RHLRHPAFVK KQHNGVCDMD CNYERFNFDG GECCDPEITN VTQTCFDPDS PHRAYLDVNE LKNILKLDGS THLNIFFAKS SEEELAGVAT WPWDKEALMH LGGIVLNPSF YGMPGHTHTM IHEIGHSLGL YHVFRGISEI QSCSDPCMET EPSFETGDLC NDTNPAPKHK SCGDPGPGND TCGFHSFFNT PYNNFMSYAD DDCTDSFTPN QVARMHCYLD LVYQGWQPSR KPAPVALAPQ VLGHTTDSVT LEWFPPIDGH FFERELGSAC HLCLEGRILV QYASNASSPM PCSPSGHWSP REAEGHPDVE QPCKSSVRTW SPNSAVNPHT VPPACPEPQG CYLELEFLYP LVPESLTIWV TFVSTDWDSS GAVNDIKLLA VSGKNISLGP QNVFCDVPLT IRLWDVGEEV YGIQIYTLDE HLEIDAAMLT STADTPLCLQ CKPLKYKVVR DPPLQMDVAS ILHLNRKFVD MDLNLGSVYQ YWVITISGTE ESEPSPAVTY IHGSGYCGDG IIQKDQGEQC DDMNKINGDG CSLFCRQEVS FNCIDEPSRC YFHDGDGVCE EFEQKTSIKD CGVYTPQGFL DQWASNASVS HQDQQCPGWV IIGQPAASQV CRTKVIDLSE GISQHAWYPC TISYPYSQLA QTTFWLRAYF SQPMVAAAVI VHLVTDGTYY GDQKQETISV QLLDTKDQSH DLGLHVLSCR NNPLIIPVVH DLSQPFYHSQ AVRVSFSSPL VAISGVALRS FDNFDPVTLS SCQRGETYSP AEQSCVHFAC EKTDCPELAV ENASLNCSSS DRYHGAQCTV SCRTGYVLQI RRDDELIKSQ TGPSVTVTCT EGKWNKQVAC EPVDCSIPDH HQVYAASFSC PEGTTFGSQC SFQCRHPAQL KGNNSLLTCM EDGLWSFPEA LCELMCLAPP PVPNADLQTA RCRENKHKVG SFCKYKCKPG YHVPGSSRKS KKRAFKTQCT QDGSWQEGAC VPVTCDPPPP KFHGLYQCTN GFQFNSECRI KCEDSDASQG LGSNVIHCRK DGTWNGSFHV CQEMQGQCSV PNELNSNLKL QCPDGYAIGS ECATSCLDHN SESIILPMNV TVRDIPHWLN PTRVERVVCT AGLKWYPHPA LIHCVKGCEP FMGDNYCDAI NNRAFCNYDG GDCCTSTVKT KKVTPFPMSC DLQGDCACRD PQAQEHSRKD LRGYSHG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali