missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PGD / Phosphogluconate dehydrogenase Protein

A DNA sequence encoding the Homo sapiens (Human) PGD / Phosphogluconate dehydrogenase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8351-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16014171

  • 2385.00€ / 1mg
Spedizione stimata: 13-06-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

NP_001291380.1
PGD / Phosphogluconate dehydrogenase Protein
Research Use Only
PGD
Recombinant Protein
E. coli
ADIALIGLAV MGQNLILNMN DHGFVVCAFN RTVSKVDDFL ANEAKGTKVV GAQSLKEMVS KLKKPRRIIL LVKAGQAVDD FIEKLVPLLD TGDIIIDGGN SEYRDTTRRC RDLKAKGILF VGSGVSGGEE GARYGPSLMP GGNKEAWPHI KTIFQGIAAK VGTGEPCCDW VGDEGAGHFV KMVHNGIEYG DMQLICEAYH LMKDVLGMAQ DEMAQAFEDW NKTELDSFLI EITANILKFQ DTDGKHLLPK IRDSAGQKGT GKWTAISALE YGVPVTLIGE AVFARCLSSL KDERIQASKK LKGPQKFQFD GDKKSFLEDI RKALYASKII SYAQGFMLLR QAATEFGWTL NYGGIALMWR GGCIIRSVFL GKIKDAFDRN PELQNLLLDD FFKSAVENCQ DSWRRAVSTG VQAGIPMPCF TTALSFYDGY RHEMLPASLI QAQRDYFGAH TYELLAKPGQ FIHTNWTGHG GTVSSSSYNA
Greater than 90% as determined by SDS-PAGE.
5226
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali