missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PI3Kb Protein
A cDNA sequence encoding the PI3Kb was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10818-1ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
PI3Kb Protein | |
Research Use Only | |
The specific activity was found to be 3,000 units/mg (1 unit is defined as 1 picomole phosphate transferred to PIP2 per minute). | |
Human | |
Untagged | |
Greater than 90.0% as determined by SDS Page. |
1 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Baculovirus | |
0.9 mg/ml solution in PBS and 2mM MgCl2. |