missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PIGX Protein

A DNA sequence encoding the Homo sapiens (Human) PIGX, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7951-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16061801

Quantità Prezzo
1 160.00€ / 10µg
10 148.99€ / 10µg
Spedizione stimata
28-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001159776.1
PIGX Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
54965
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PIGX
Recombinant Protein
E. coli
MCSEIILRQE VLKDGFHRDL LIKVKFGESI EDLHTCRLLI KQDIPAGLYV DPYELASLRE RNITEAVMVS ENFDIEAPNY LSKESEVLIY ARRDSQCIDC FQAFLPVHCR YHRPHSEDGE ASIVVNNPDL LMFCDQEFPI LKCWAHSEVA APCALENEDI CQWNKMKYKS VYKNVILQVP VGLTVHTSL
Videos
Documenti