missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP3CC Protein
A DNA sequence encoding the Homo sapiens (Human) PPP3CC, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6523-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001230903.1 | |
PPP3CC Protein | |
Research Use Only | |
PPP3CC | |
Recombinant Protein | |
E. coli | |
SGRRFHLSTT DRVIKAVPFP PTQRLTFKEV FENGKPKVDV LKNHLVKEGR LEEEVALKII NDGAAILRQE KTMIEVDAPI TVCGDIHGQF FDLMKLFEVG GSPSNTRYLF LGDYVDRGYF SIECVLYLWS LKINHPKTLF LLRGNHECRH LTDYFTFKQE CRIKYSEQVY DACMETFDCL PLAALLNQQF LCVHGGMSPE ITSLDDIRKL DRFTEPPAFG PVCDLLWSDP SEDYGNEKTL EHYTHNTVRG CSYFYSYPAV CEFLQNNNLL SIIRAHEAQD AGYRMYRKSQ ATGFPSLITI FSAPNYLDVY NNKAAVLKYE NNVMNIRQFN CSPHPYWLPN FMDVFTWSLP FVGEKVTEML VNVLNICSDD ELISDDEAEG STTVRKEIIR NKIRAIGKMA RVFSILRQES ESVLTLKGLT PTGTLPLGVL SGGKQTIETA TVEAVEAREA IRGFSLQHKI RSFEEARGLD RINERMPPRK DSIHAGGPMK SVTSAHSHAA HRSDQGKKAH S | |
Greater than 90% as determined by SDS-PAGE. |
5533 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |