missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit beta type-10 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6544-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002792.1 | |
Proteasome subunit beta type-10 Protein | |
Research Use Only | |
PSMB10 | |
Recombinant Protein | |
E. coli | |
TTIAGLVFQD GVILGADTRA TNDSVVADKS CEKIHFIAPK IYCCGAGVAA DAEMTTRMVA SKMELHALST GREPRVATVT RILRQTLFRY QGHVGASLIV GGVDLTGPQL YGVHPHGSYS RLPFTALGSG QDAALAVLED RFQPNMTLEA AQGLLVEAVT AGILGDLGSG GNVDACVITK TGAKLLRTLS SPTEPVKRSG RYHFVPGTTA VLTQTVKPLT LELVEETVQA MEVE | |
Greater than 80% as determined by SDS-PAGE. |
5699 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |