missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSME3 / PA28-gamma Protein
A DNA sequence encoding the Homo sapiens (Human) PSME3 / PA28-gamma, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8770-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005780.2 | |
PSME3 / PA28-gamma Protein | |
Research Use Only | |
PSME3 | |
Recombinant Protein | |
E. coli | |
ASLLKVDQEV KLKVDSFRER ITSEAEDLVA NFFPKKLLEL DSFLKEPILN IHDLTQIHSD MNLPVPDPIL LTNSHDGLDG PTYKKRRLDE CEEAFQGTKV FVMPNGMLKS NQQLVDIIEK VKPEIRLLIE KCNTVKMWVQ LLIPRIEDGN NFGVSIQEET VAELRTVESE AASYLDQISR YYITRAKLVS KIAKYPHVED YRRTVTEIDE KEYISLRLII SELRNQYVTL HDMILKNIEK IKRPRSSNAE T | |
Greater than 90% as determined by SDS-PAGE. |
10197 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |