missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPN6 Protein

A DNA sequence encoding the Homo sapiens (Human) PTPN6, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6555-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16014120

Quantità Prezzo
1 434.28€ / 100µg
6 408.22€ / 100µg
Spedizione stimata
07-06-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_002822.2
PTPN6 Protein
100 μg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
5777
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PTPN6
Recombinant Protein
E. coli
MLSRGVGDQV THIRIQNSGD FYDLYGGEKF ATLTELVEYY TQQQGVLQDR DGTIIHLKYP LNCSDPTSER WYHGHMSGGQ AETLLQAKGE PWTFLVRESL SQPGDFVLSV LSDQPKAGPG SPLRVTHIKV MCEGGRYTVG GLETFDSLTD LVEHFKKTGI EEASGAFVYL RQPYYATRVN AADIENRVLE LNKKQESEDT AKAGFWEEFE SLQKQEVKNL HQRLEGQRPE NKGKNRYKNI LPFDHSRVIL QGRDSNIPGS DYINANYIKN QLLGPDENAK TYIASQGCLE ATVNDFWQMA WQENSRVIVM TTREVEKGRN KCVPYWPEVG MQRAYGPYSV TNCGEHDTTE YKLRTLQVSP LDNGDLIREI WHYQYLSWPD HGVPSEPGGV LSFLDQINQR QESLPHAGPI IVHCSAGIGR TGTIIVIDML MENISTKGLD CDIDIQKTIQ MVRAQRSGMV QTEAQYKFIY VAIAQFIETT KKKLEVLQSQ KGQESEYGNI TYPPAMKNAH AKASRTSSKH KEDVYENLHT KNKREEKVKK QRSADKEKSK GSLKRK
Videos
SDS
Documenti