missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein Rab-18 Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-18, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8149-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062911

Quantità Prezzo
1 160.00€ / 10µg
10 148.99€ / 10µg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_001243339.1
Ras-related protein Rab-18 Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
22931
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RAB18
Recombinant Protein
E. coli
MDEDVLTTLK ILIIGESGVG KSSLLLRFTD DTFDPELAAT IGVDFKVKTI SVDGNKAKLA IWDTAGQERF RTLTPSYYRG AQGVILVYDV TRRDTFVKLD NWLNELETYC TRNDIVNMLV GNKIDKENRE VDRNEGLKFA RKHSMLFIEA SAKTCDGVQC AFEELVEKII QTPGLWESEN QNKGVKLSHR EEGQGGGACG GYC
Videos
Documenti