missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulatory protein E2 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulatory protein E2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7368-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Regulatory protein E2 Protein | |
Research Use Only | |
E2 | |
Recombinant Protein | |
Yeast | |
MQTPKETLSE RLSCVQDKII DHYENDSKDI DSQIQYWQLI RWENAIFFAA REHGIQTLNH QVVPAYNISK SKAHKAIELQ MALQGLAQSA YKTEDWTLQD TCEELWNTEP THCFKKGGQT VQVYFDGNKD NCMTYVAWDS VYYMTDAGTW DKTATCVSHR GLYYVKEGYN TFYIEFKSEC EKYGNTGTWE VHFGNNVIDC NDSMCSTSDD TVSATQLVKQ LQHTPSPYSS TVSVGTAKTY GQTSAATRPG HCGLAEKQHC GPVNPLLGAA TPTGNNKRRK LCSGNTTPII HLKGDRNSLK CLRYRLRKHS DHYRDISSTW HWTGAGNEKT GILTVTYHSE TQRTKFLNTV AIPDSVQILV GYMTM | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |