missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Replication protein A 32 kDa subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Replication protein A 32 kDa subunit, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8374-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001273005.1 | |
Replication protein A 32 kDa subunit Protein | |
Research Use Only | |
RPA2 | |
Recombinant Protein | |
E. coli | |
MWNSGFESYG SSSYGGAGGY TQSPGGFGSP APSQAEKKSR ARAQHIVPCT ISQLLSATLV DEVFRIGNVE ISQVTIVGII RHAEKAPTNI VYKIDDMTAA PMDVRQWVDT DDTSSENTVV PPETYVKVAG HLRSFQNKKS LVAFKIMPLE DMNEFTTHIL EVINAHMVLS KANSQPSAGR APISNPGMSE AGNFGGNSFM PANGLTVAQN QVLNLIKACP RPEGLNFQDL KNQLKHMSVS SIKQAVDFLS NEGHIYSTVD DDHFKST | |
Greater than 90% as determined by SDS-PAGE. |
6118 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |