missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinol dehydrogenase 11 Protein
A DNA sequence encoding the Homo sapiens (Human) Retinol dehydrogenase 11, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6592-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001239579.1 | |
Retinol dehydrogenase 11 Protein | |
Research Use Only | |
RDH11 | |
Recombinant Protein | |
E. coli | |
PQIRKMLSSG VCTSTVQLPG KVVVVTGANT GIGKETAKEL AQRGARVYLA CRDVEKGELV AKEIQTTTGN QQVLVRKLDL SDTKSIRAFA KGFLAEEKHL HVLINNAGVM MCPYSKTADG FEMHIGVNHL GHFLLTHLLL EKLKESAPSR IVNVSSLAHH LGRIHFHNLQ GEKFYNAGLA YCHSKLANIL FTQELARRLK GSGVTTYSVH PGTVQSELVR HSSFMRWMWW LFSFFIKTPQ QGAQTSLHCA LTEGLEILSG NHFSDCHVAW VSAQARNETI ARRLWDVSCD LLGLPID | |
Greater than 90% as determined by SDS-PAGE. |
51109 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |