missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ropporin-1B Protein
A DNA sequence encoding the Homo sapiens (Human) Ropporin-1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6614-ye-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001012337.1 | |
Ropporin-1B Protein | |
Research Use Only | |
ROPN1B | |
Recombinant Protein | |
Yeast | |
MWKVVNLPTD LFNSVMNVGR FTEEIEWLKF LALACSALGV TITKTLKIVC EVLSCDHNGG LPRIPFSTFQ FLYTYIAEVD GEICASHVSR MLNYIEQEVI GPDGLITVND FTQNPRVWLE | |
Greater than 90% as determined by SDS-PAGE. |
152015 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |