missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RPA1 Protein

A DNA sequence encoding the Homo sapiens (Human) RPA1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6618-ec-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16064590

Quantità Prezzo
1 1180.00€ / 500µg
3 1100.47€ / 500µg
Spedizione stimata
21-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_002936.1
RPA1 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
6117
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPA1
Recombinant Protein
E. coli
VGQLSEGAIA AIMQKGDTNI KPILQVINIR PITTGNSPPR YRLLMSDGLN TLSSFMLATQ LNPLVEEEQL SSNCVCQIHR FIVNTLKDGR RVVILMELEV LKSAEAVGVK IGNPVPYNEG LGQPQVAPPA PAASPAASSR PQPQNGSSGM GSTVSKAYGA SKTFGKAAGP SLSHTSGGTQ SKVVPIASLT PYQSKWTICA RVTNKSQIRT WSNSRGEGKL FSLELVDESG EIRATAFNEQ VDKFFPLIEV NKVYYFSKGT LKIANKQFTA VKNDYEMTFN NETSVMPCED DHHLPTVQFD FTGIDDLENK SKDSLVDIIG ICKSYEDATK ITVRSNNREV AKRNIYLMDT SGKVVTATLW GEDADKFDGS RQPVLAIKGA RVSDFGGRSL SVLSSSTIIA NPDIPEAYKL RGWFDAEGQA LDGVSISDLK SGGVGGSNTN WKTLYEVKSE NLGQGDKPDY FSSVATVVYL RKENCMYQAC PTQDCNKKVI DQQNGLYRCE KCDTEFPNFK YRMILSVNIA DFQENQWVTC FQESAEAILG QNAAYLGELK DKNEQAFEEV FQNANFRSFI FRVRVKVETY NDESRIKATV MDVKPVDYRE YGRRLVMSIR RSALM
Videos
Documenti