missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS3A Protein
A cDNA sequence encoding the RPS3A was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13357-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
RPS3A Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RPS3A solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 50% glycerol. |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAVGKN KRLTKGGKKG AKKKVVDPFS KKDWYDVKAP AMFNIRNIGK TLVTRTQGTK IASDGLKGRV FEVSLADLQN DEVAFRKFKL ITEDVQGKNC LTNFHGMDLT RDKMCSMVKK WQTMIEAHVD VKTTDGYLLR LFCVGFTKKR NNQIRKTSYA QHQQVRQIRK KMMEIMTREV QTNDLKEVVN KLIPDSIGKD IEKACQSIYP LHDVFVRKVK MLKKPKFELG KLMELHGEGS SSGKATGDET GAKVERADGY EPPVQESV |